Proteintech
Manufacturer Code:665611IG150UL
Catalog # 50-173-6980
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 2E6B6 |
Immunogen | SMARCA4 Fusion Protein Ag16256 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: ATP dependent helicase SMARCA4 BAF190 BAF190A BRG1 BRG1 associated factor 190A hSNF2b Protein brahma homolog 1 Protein BRG 1 SMARCA4 SNF2 SNF2 BETA SNF2B SNF2L4 SNF2LB SWI2 Transcription activator BRG1; ATP-dependent helicase SMARCA4; BAF190A; brahma protein-like 1; BRG1-associated factor 190A; BRM/SWI2-related gene 1; global transcription activator homologous sequence; homeotic gene regulator; Mitotic growth and transcription activator; nuclear protein GRB1; Protein brahma homolog 1; Protein BRG-1; SNF2-beta; SNF2-like 4; sucrose nonfermenting-like 4; SWI/SNF-related matrix-associated actin-dependent regulator of chromatin subfamily A member 4; Transcription activator BRG1
Gene Aliases: BAF190; BAF190A; brg-1; BRG1; CSS4; hSNF2b; MRD16; RTPS2; SMARCA4; SNF2; SNF2B; SNF2L4; SNF2LB; SWI2
UniProt ID: (Human) P51532
Entrez Gene ID: (Human) 6597, (Rat) 171379
Molecular Function: DNA binding protein DNA helicase helicase nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.