Invitrogen
Manufacturer Code:PA583119
Catalog # PIPA583119
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human SMARCA2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ATP-dependent helicase SMARCA2; BAF190 BRM hBRM hSNF2a SNF2 SNF2L2 SNF2LA Sth1p SWI2; BAF190B; BRG1-associated factor 190B; global transcription activator homologous sequence; hBRM; Probable global transcription activator SNF2L2; Protein brahma homolog; SNF2-alpha; SNF2/SWI2-like protein 2; sucrose nonfermenting 2-like protein 2; SWI/SNF-related matrix-associated actin-dependent regulator of chromatin a2; SWI/SNF-related matrix-associated actin-dependent regulator of chromatin subfamily A member 2
Gene Aliases: BAF190; BAF190B; BRM; hBRM; hSNF2a; NCBRS; SMARCA2; SNF2; SNF2A; SNF2L2; SNF2LA; Sth1p; SWI2
UniProt ID: (Human) P51531
Entrez Gene ID: (Human) 6595
Molecular Function: DNA binding protein DNA helicase helicase nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.