Invitrogen
Manufacturer Code:PA5103113
Catalog # PIPA5103113
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human Brm corresponding to a region within C-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2610209L14Rik ATP-dependent helicase SMARCA2 BAF190 Baf190b brahma BRG1-associated factor 190B BRM FLJ36757 global transcription activator homologous sequence hBRM hSNF2a MGC74511 NCBRS Probable global transcription activator SNF2L2 protein brahma homolog SMARCA2 SNF2 SNF2/SWI2-like protein 2 SNF2A SNF2alpha SNF2-alpha Snf2l2 SNF2LA Sth1p subunit of the SWI/SNF chromatin remodeling complex sucrose nonfermenting 2-like protein 2 SWI/SNF related matrix associated actin dependent regulator of chromatin subfamily a member 2 SWI/SNF-related matrix-associated actin-dependent regulator of chromatin a2 SWI/SNF-related matrix-associated actin-dependent regulator of chromatin subfamily A member 2 SWI2; ATP-dependent helicase SMARCA2; BAF190B; BRG1-associated factor 190B; global transcription activator homologous sequence; hBRM; Probable global transcription activator SNF2L2; Protein brahma homolog; SNF2-alpha; SNF2/SWI2-like protein 2; subunit of the SWI/SNF chromatin remodeling complex; sucrose nonfermenting 2-like protein 2; SWI/SNF-related matrix-associated actin-dependent regulator of chromatin a2; SWI/SNF-related matrix-associated actin-dependent regulator of chromatin subfamily A member 2
Gene Aliases: 2610209L14Rik; BAF190; BAF190B; brahma; BRM; hBRM; hSNF2a; NCBRS; SMARCA2; SNF2; SNF2A; SNF2alpha; SNF2L2; SNF2LA; Sth1p; SWI2
UniProt ID: (Human) P51531, (Mouse) Q6DIC0
Entrez Gene ID: (Human) 6595, (Mouse) 67155, (Rat) 361745
Molecular Function:
DNA binding protein
DNA helicase
helicase
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.