Invitrogen
Manufacturer Code:PA568876
Catalog # PIPA568876
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ChIP assay (ChIP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the N terminal region of mouse SMARCA1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ATP-dependent helicase SMARCA1; DNA-dependent ATPase SNF2L; Nucleosome-remodeling factor subunit SNF2L; Probable global transcription activator SNF2L1; RP3-353H6.2 DKFZp686D1623 FLJ41547 ISWI NURF140 SNF2L SNF2L1 SNF2LB SNF2LT SWI SWI2 ATP-dependent helicase SMARCA1 SNF2-like 1 SWI/SNF-related matrix-associated actin-dependent regulator of chromatin a1 SWI/SNF-related matrix-associated actin-dependent regulator of chromatin subfamily A member 1 global transcription activator homologous sequence nucleosome-remodeling factor subunit SNF2L probable global transcription activator SNF2L1 sucrose nonfermenting 2-like protein 1; SWI/SNF-related matrix-associated actin-dependent regulator of chromatin subfamily A member 1
Gene Aliases: 5730494M04Rik; RGD1561046; Smarca1; Snf2l
UniProt ID: (Mouse) Q6PGB8
Entrez Gene ID: (Rat) 317575, (Mouse) 93761
Molecular Function: DNA binding protein DNA helicase helicase nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.