Invitrogen
Manufacturer Code:PA540911
Catalog # PIPA540911
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human SMARCA1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ATP-dependent helicase SMARCA1; DNA-dependent ATPase SNF2L; global transcription activator homologous sequence; Nucleosome-remodeling factor subunit SNF2L; Probable global transcription activator SNF2L1; SNF2-like 1; SNF2L-like protein; sucrose nonfermenting 2-like protein 1; SWI/SNF related matrix associated actin dependent regulator of Chromatin subfamily a member 1 DKFZp686D1623 FLJ41547 ISWI NURF140 SNF2L SNF2L1 SNF2LB SWI SWI2 SNF2LT; SWI/SNF-related matrix-associated actin-dependent regulator of chromatin subfamily A member 1
Gene Aliases: 5730494M04Rik; hSNF2L; ISWI; NURF140; RGD1561046; SMARCA1; SNF2L; SNF2L1; SNF2LB; SNF2LT; SWI; SWI2
UniProt ID: (Human) P28370, (Mouse) Q6PGB8
Entrez Gene ID: (Dog) 481046, (Human) 6594, (Bovine) 535439, (Horse) 100054822, (Pig) 100188905, (Rabbit) 100345455, (Rat) 317575, (Mouse) 93761
Molecular Function:
DNA binding protein
DNA helicase
helicase
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.