Invitrogen
Manufacturer Code:PA5111587
Catalog # PIPA5111587
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant Protein corresonding to Human SMARCA1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 5730494M04Rik ATP-dependent helicase SMARCA1 BOS_25187 DKFZp686D1623 DNA-dependent ATPase SNF2L FLJ41547 hSNF2L ISWI Nucleosome-remodeling factor subunit SNF2L NURF140 Probable global transcription activator SNF2L1 RGD1561046 RP3-353H6.2 Smarca1 SNF2L SNF2L1 SNF2LB SNF2LT SWI SWI/SNF-related matrix-associated actin-dependent regulator of chromatin subfamily A member 1 SWI2; ATP-dependent helicase SMARCA1; global transcription activator homologous sequence; Nucleosome-remodeling factor subunit SNF2L; Probable global transcription activator SNF2L1; SNF2-like 1; sucrose nonfermenting 2-like protein 1; SWI/SNF-related matrix-associated actin-dependent regulator of chromatin subfamily A member 1
Gene Aliases: hSNF2L; ISWI; NURF140; SMARCA1; SNF2L; SNF2L1; SNF2LB; SNF2LT; SWI; SWI2
UniProt ID: (Human) P28370
Entrez Gene ID: (Human) 6594
Molecular Function:
DNA binding protein
DNA helicase
helicase
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.