Novus Biologicals
Manufacturer Code:NBP183087
Catalog # NBP183087
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids:EQGSRVLIFSQMTRLLDILEDYCMWRGYEYCRLDGQTPHEEREDKFLEVEFLGQREAIEAFNAPNSSKFIFMLSTRAGGLGINLASADVVILYDSDWNPQVD |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ATP-dependent helicase SMARCA1; ATP-dependent helicase SMARCA1 DKFZp686D1623 EC 3.6.1 EC 3.6.4.- FLJ41547 global transcription activator homologous sequence Nucleosome-remodeling factor subunit SNF2L NURF140 probable global transcription activator SNF2L1 SNF2L SNF2L1ISWI SNF2LB SNF2-like 1 SNF2LT subfamily a member 1 sucrose nonfermenting 2-like protein 1 SWI SWI/SNF related matrix associated actin dependent regulator of chromatin SWI/SNF-related matrix-associated actin-dependent regulator of chromatin a1 SWI/SNF-related matrix-associated actin-dependent regulator of chromatinsubfamily A member 1 SWI2; global transcription activator homologous sequence; Nucleosome-remodeling factor subunit SNF2L; Probable global transcription activator SNF2L1; SNF2-like 1; sucrose nonfermenting 2-like protein 1; SWI/SNF-related matrix-associated actin-dependent regulator of chromatin subfamily A member 1
Gene Aliases: hSNF2L; ISWI; NURF140; SMARCA1; SNF2L; SNF2L1; SNF2LB; SNF2LT; SWI; SWI2
UniProt ID: (Human) P28370
Entrez Gene ID: (Human) 6594
Molecular Function:
DNA binding protein
DNA helicase
helicase
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.