Proteintech
Manufacturer Code:163971AP150UL
Catalog # 50-173-0920
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | SMAD9 Fusion Protein Ag9490 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: MAD homolog 9; MAD homolog 9 MADH6 MADH9 Mothers against DPP homolog 9 SMAD 9 SMAD family member 9 SMAD8A SMAD8B SMAD9; Madh6; Mothers against decapentaplegic homolog 9; Mothers against decapentaplegic, drosophila, homolog of, 9; mothers against DPP homolog 9; SMAD 9; SMAD family member 9; SMAD, mothers against DPP homolog 9; Smad8
Gene Aliases: MADH6; Madh8; MADH9; PPH2; SMAD8; SMAD8/9; SMAD8A; SMAD8B; SMAD9
UniProt ID: (Human) O15198, (Rat) O54835, (Mouse) Q9JIW5
Entrez Gene ID: (Human) 4093, (Rat) 85435, (Mouse) 55994
Molecular Function:
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.