Proteintech
Manufacturer Code:664781IG150UL
Catalog # 50-173-6901
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 2B9A4 |
Immunogen | SMAD7 Fusion Protein Ag13688 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: CRCS3 hSMAD7 MAD homolog 7 MAD homolog 8 MADH7 MADH8 Mothers against DPP homolog 7 Mothers against DPP homolog 8 SMAD 7 SMAD family member 7 SMAD7; hSMAD7; MAD (mothers against decapentaplegic, Drosophila) homolog 7; MAD homolog 7; MAD homolog 8; madh8; Mothers against decapentaplegic homolog 7; Mothers against decapentaplegic homolog 8; Mothers against decapentaplegic, drosophila, homolog of, 7; mothers against decapentaplegic-like protein 7; mothers against DPP 7; mothers against DPP homolog 7; mothers against DPP homolog 8; SMAD 7; SMAD family member 7; SMAD, mothers against DPP homolog 7
Gene Aliases: CRCS3; MADH7; MADH8; SMAD7
UniProt ID: (Human) O15105, (Mouse) O35253, (Rat) O88406
Entrez Gene ID: (Pig) 100521305, (Human) 4092, (Mouse) 17131, (Rat) 81516
Molecular Function: transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.