Invitrogen
Manufacturer Code:PA129829
Catalog # PIPA129829
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptides RAGGPPRPMSEPGAG and NLEQRSESVRRTRSKIG corresponding to amino acids 85-99 and 372-388 of Human SMAD6. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: HsT17432 MADH6 MADH7; MAD homolog 6; Mad homolog 7; Mothers against decapentaplegic homolog 6; mothers against decapentaplegic, drosophila, homolog of, 6; mothers against DPP homolog 6; SMAD 6; SMAD family member 6; SMAD, mothers against DPP homolog 6
Gene Aliases: AOVD2; b2b390Clo; HsT17432; MADH6; MADH7; Msmad6; SMAD6
UniProt ID: (Human) O43541, (Mouse) O35182
Entrez Gene ID: (Human) 4091, (Mouse) 17130, (Rat) 367100
Molecular Function: transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.