Abnova
Manufacturer Code:PAB0368
Catalog # 89-114-180
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A mixture of synthetic peptides corresponding to amino acids 85-99 and 372-388 of human SMAD6 . |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of SMAD6.
For Research Use Only
Protein Aliases: HsT17432; MAD homolog 6; MADH6; MADH7; Mothers against decapentaplegic homolog 6; mothers against decapentaplegic, drosophila, homolog of, 6; SMAD 6; SMAD family member 6; SMAD, mothers against DPP homolog 6
Gene Aliases: AOVD2; HsT17432; MADH6; MADH7; SMAD6
UniProt ID: (Human) O43541
Entrez Gene ID: (Human) 4091
Molecular Function:
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.