Proteintech
Manufacturer Code:121671AP150UL
Catalog # 50-172-7763
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | SMAD5 Fusion Protein Ag2809 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Dwarfin-C; Dwf-C; Dwfc hSmad5 JV5 1 MAD homolog 5 MADH5 Mothers against DPP homolog 5 SMAD 5 SMAD family member 5 SMAD5; JV5-1; MAD homolog 5; MAD, mothers against decapentaplegic homolog 5; Mothers against decapentaplegic homolog 5; mothers against decapentaplegic, drosophila, homolog of, 5; mothers against DPP homolog 5; mSmad5; SMA- and MAD-related protein 5; SMAD 5; SMAD family member 5; SMAD, mothers against DPP homolog 5
Gene Aliases: 1110051M15Rik; AI451355; Dwf-C; DWFC; JV5-1; MADH5; Msmad5; MusMLP; SMAD5
UniProt ID: (Human) Q99717, (Mouse) P97454, (Rat) Q9R1V3
Entrez Gene ID: (Human) 4090, (Mouse) 17129, (Rat) 59328
Molecular Function:
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.