Invitrogen
Manufacturer Code:PA131458
Catalog # PIPA131458
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to amino acids CHTMPIADPQPLD from C-Terminus of Human MADH4 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: deleted in pancreatic carcinoma locus 4; Deletion target in pancreatic carcinoma 4; Deletion target in pancreatic carcinoma 4 homolog; DPC4 JIP MADH4; MAD homolog 4; Mothers against decapentaplegic homolog 4; mothers against decapentaplegic, Drosophila, homolog of, 4; mothers against DPP homolog 4; SMAD 4; SMAD family member 4; SMAD, mothers against DPP homolog 4
Gene Aliases: AW743858; D18Wsu70e; DPC4; JIP; MADH4; MYHRS; SMAD4
UniProt ID: (Human) Q13485, (Mouse) P97471
Entrez Gene ID: (Human) 4089, (Mouse) 17128
Molecular Function:
nucleic acid binding
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.