Abnova
Manufacturer Code:PAB7689
Catalog # 89-133-427
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to residues surrounding S467 of human SMAD2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of SMAD2.
For Research Use Only
Protein Aliases: hMAD-2; hSMAD2; JV18; JV18-1; MAD homolog 2; Mad-related protein 2; MADH2; MADR2; MGC22139; MGC34440; mother against DPP homolog 2; Mothers against decapentaplegic homolog 2; Sma- and Mad-related protein 2; SMAD 2; SMAD family member 2; SMAD, mothers against DPP homolog 2
Gene Aliases: hMAD-2; hSMAD2; JV18; JV18-1; MADH2; MADR2; SMAD2
UniProt ID: (Human) Q15796
Entrez Gene ID: (Human) 4087
Molecular Function: transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.