Abnova
Manufacturer Code:H00004086M03A
Catalog # 89-104-868
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 2E9 |
Immunogen | SMAD1 (AAH01878.11 a.a. &bsim 465 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against a full-length recombinant SMAD1.
For Research Use Only
Protein Aliases: BSP-1; BSP1; JV4-1; JV41; MAD homolog 1; MAD, mothers against decapentaplegic homolog 1; Mad-related protein 1; MADH1; MADR1; Mothers against decapentaplegic homolog 1; mothers against DPP homolog 1; SMAD 1; SMAD family member 1; SMAD, mothers against DPP homolog 1; TGF-beta signaling protein 1; transforming growth factor-beta signaling protein 1; Transforming growth factor-beta-signaling protein 1
Gene Aliases: BSP-1; BSP1; JV4-1; JV41; MADH1; MADR1; SMAD1
UniProt ID: (Human) Q15797
Entrez Gene ID: (Human) 4086
Molecular Function: transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.