Invitrogen
Manufacturer Code:701168
Catalog # PI701168
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ChIP assay (ChIP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | 12H1L14 |
Immunogen | Peptide corresponding to amino acids 178193 of human SMAD1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Recombinant Monoclonal Antibody
For Research Use Only
Protein Aliases: BSP-1; BSP-1 MAD homolog 1 MAD mothers against decapentaplegic homolog 1 Mad-related protein 1 TGF-beta signaling protein 1 hSMAD1 mothers against DPP homolog 1 transforming growth factor-beta signaling protein 1 SMAD1 DWFC JV5-1 MADH5 SMAD5; Dwarfin-A; Dwf-A; JV4-1; MAD homolog 1; MAD, mothers against decapentaplegic homolog 1; Mad-related protein 1; mMad1; Mothers against decapentaplegic homolog 1; mothers against DPP homolog 1; Mothers-against-DPP-related 1; SMAD 1; SMAD family member 1; SMAD, mothers against DPP homolog 1; TGF-beta signaling protein 1; transforming growth factor-beta signaling protein 1; Transforming growth factor-beta-signaling protein 1
Gene Aliases: AI528653; BSP-1; BSP1; JV4-1; JV41; MADH1; MADR1; Mlp1; MusMLP; SMAD1
UniProt ID: (Human) Q15797, (Mouse) P70340
Entrez Gene ID: (Human) 4086, (Mouse) 17125
Molecular Function: transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.