Proteintech
Manufacturer Code:104291AP150UL
Catalog # 50-172-6302
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | SMAD1 Fusion Protein Ag0701 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: BSP 1 BSP1 hSMAD1 JV4 1 JV41 MAD homolog 1 Mad related protein 1 MADH1 MADR1 Mothers against DPP homolog 1 SMAD 1 SMAD family member 1 SMAD1; BSP-1; Dwarfin-A; Dwf-A; JV4-1; MAD (mothers against decapentaplegic, Drosophila) homolog 1; MAD homolog 1; MAD homolog1 (mothers against decapentaplegic, Drosophila); MAD, mothers against decapentaplegic homolog 1; Mad-related protein 1; mMad1; Mothers against decapentaplegic homolog 1; mothers against DPP homolog 1; Mothers-against-DPP-related 1; SMAD 1; SMAD family member 1; SMAD, mothers against DPP homolog 1; TGF-beta signaling protein 1; transforming growth factor-beta signaling protein 1; Transforming growth factor-beta-signaling protein 1
Gene Aliases: AI528653; BSP-1; BSP1; JV4-1; JV41; Mad1; MADH1; MADR1; Mlp1; MusMLP; SMAD1
UniProt ID: (Human) Q15797, (Mouse) P70340, (Rat) P97588
Entrez Gene ID: (Human) 4086, (Mouse) 17125, (Rat) 25671
Molecular Function:
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.