Novus Biologicals
Manufacturer Code:NBP267768
Catalog # NBP267768
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | SD2090 |
Immunogen | Recombinant Protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Rabbit Monoclonal Antibody
Protein Aliases: BSP-1; BSP1 BSP-1 hSMAD1 JV41 JV4-1 JV4-1SMAD mothers against DPP homolog 1 (Drosophila) MAD homolog 1 MADH1 MADH1JV41 MADR1 MADR1MAD mothers against decapentaplegic homolog 1 (Drosophila) Mad-related protein 1 mothers against decapentaplegic homolog 1 Mothers against DPP homolog 1 SMAD family member 1 SMAD family member 1SMAD 1 SMAD mothers against DPP homolog 1 Smad1 TGF-beta signaling protein 1 transforming growth factor-beta signaling protein 1; JV4-1; MAD homolog 1; MAD, mothers against decapentaplegic homolog 1; Mad-related protein 1; Mothers against decapentaplegic homolog 1; mothers against DPP homolog 1; SMAD 1; SMAD family member 1; SMAD, mothers against DPP homolog 1; TGF-beta signaling protein 1; transforming growth factor-beta signaling protein 1; Transforming growth factor-beta-signaling protein 1
Gene Aliases: BSP-1; BSP1; JV4-1; JV41; MADH1; MADR1; SMAD1
UniProt ID: (Human) Q15797
Entrez Gene ID: (Human) 4086
Molecular Function: transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.