Abnova
Manufacturer Code:H00004092M06
Catalog # 89-000-789
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 4E1 |
Immunogen | SMAD7 (NP_005895 160 a.a. ~ 260 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against a partial recombinant SMAD7.
For Research Use Only
Protein Aliases: CRCS3; FLJ16482; hSMAD7; MAD (mothers against decapentaplegic, Drosophila) homolog 7; MAD homolog 7; MAD homolog 8; MADH7; MADH8; Mothers against decapentaplegic homolog 7; Mothers against decapentaplegic homolog 8; Mothers against decapentaplegic, drosophila, homolog of, 7; mothers against DPP homolog 8; SMAD 7; SMAD family member 7; SMAD, mothers against DPP homolog 7
Gene Aliases: CRCS3; MADH7; MADH8; SMAD7
UniProt ID: (Human) O15105
Entrez Gene ID: (Human) 4092
Molecular Function:
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.