Invitrogen
Manufacturer Code:PA546622
Catalog # PIPA546622
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the C-terminal of human SLX1B |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: GIY-YIG domain-containing protein 1; SLX1 homolog B structure-specific endonuclease subunit; SLX1 structure-specific endonuclease subunit homolog A; SLX1A; SLX1B; Structure-specific endonuclease subunit SLX1
Gene Aliases: GIYD1; GIYD2; SLX1; SLX1A; SLX1B
UniProt ID: (Human) Q9BQ83
Entrez Gene ID: (Horse) 100066262, (Human) 548593
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.