Novus Biologicals
Manufacturer Code:NBP15685920UL
Catalog # NBP15685920
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptides corresponding to SLMO2(slowmo homolog 2 (Drosophila)) The peptide sequence was selected from the N terminal of SLMO2. Peptide sequence GVDVLDRHIDPSGKLHSHRLLSTEWGLPSIVKSLIGAARTKTYVQEHSVV. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C20orf45 chromosome 20 open reading frame 45 dJ543J19.5 PRELID3B protein slowmo homolog 2 slowmo homolog 2 (Drosophila); PRELI domain containing protein 3B; Protein slowmo homolog 2; slowmo homolog 2
Gene Aliases: C20orf45; CGI-107; dJ543J19.5; PRELID3B; SLMO2
UniProt ID: (Human) Q9Y3B1
Entrez Gene ID: (Human) 51012
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.