Proteintech
Manufacturer Code:211501AP150UL
Catalog # 50-173-3283
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | SLITRK6 Fusion Protein Ag15701 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 4832410J21Rik; neuronal transmembrane protein Slitrk6; SLIT and NTRK like protein 6 SLITRK6; SLIT and NTRK-like family, member 6; SLIT and NTRK-like protein 6; slit and trk like gene 6
Gene Aliases: 4832410J21Rik; DFNMYP; SLITRK6; Sltk6
UniProt ID: (Human) Q9H5Y7, (Mouse) Q8C110
Entrez Gene ID: (Human) 84189, (Mouse) 239250, (Rat) 290467
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.