Abnova
Manufacturer Code:H00006586M07
Catalog # 89-123-291
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 1D11 |
Immunogen | SLIT3 (NP_003053 1371 a.a. ~ 1470 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against a partial recombinant SLIT3.
For Research Use Only
Protein Aliases: FLJ10764; MEGF5; Multiple EGF-like domains protein 5; Multiple epidermal growth factor-like domains protein 5; SLIL2; slit homolog 3; Slit homolog 3 protein; Slit-3; SLIT1; slit2
Gene Aliases: KIAA0814; MEGF5; SLIL2; Slit-3; SLIT1; slit2; SLIT3; UNQ691/PRO1336
UniProt ID: (Human) O95804
Entrez Gene ID: (Human) 6586
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.