Invitrogen
Manufacturer Code:PA544935
Catalog # PIPA544935
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human SLIT3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Multiple EGF-like domains protein 5; multiple epidermal growth factor-like domains 5; Multiple epidermal growth factor-like domains protein 5; slit homolog 3; Slit homolog 3 protein; Slit-3
Gene Aliases: b2b2362.1Clo; KIAA0814; MEGF5; SLIL2; Slit-3; SLIT1; slit2; SLIT3; UNQ691/PRO1336
UniProt ID: (Human) O95804, (Rat) O88280, (Mouse) Q9WVB4
Entrez Gene ID: (Dog) 489132, (Bovine) 615883, (Rabbit) 100338309, (Horse) 100070488, (Human) 6586, (Rat) 83467, (Guinea pig) 100713481, (Mouse) 20564
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.