Invitrogen
Manufacturer Code:PA146148
Catalog # PIPA146148
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide made to C-terminal region of Human SLIRP protein. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DC50 PD04872 SLIRP; SRA stem-loop-interacting RNA-binding protein, mitochondrial
Gene Aliases: 1810035L17Rik; C14orf156; DC23; DC50; PD04872; SLIRP
UniProt ID: (Human) Q9GZT3, (Mouse) Q9D8T7
Entrez Gene ID: (Human) 81892, (Mouse) 380773
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.