Novus Biologicals
Manufacturer Code:NB11037258R
Catalog # NB11037258R
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide made to a C-terminal region of human SLIRP protein. [Entrez-Prot# AAX58600] |
Conjugate | DyLight 550 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: chromosome 14 open reading frame 156 DC50 PD04872 SLIRP SRA stem-loop-interacting RNA-binding protein mitochondrial; SRA stem-loop-interacting RNA-binding protein, mitochondrial
Gene Aliases: C14orf156; DC23; DC50; PD04872; SLIRP
UniProt ID: (Human) Q9GZT3
Entrez Gene ID: (Human) 81892
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.