Abnova
Manufacturer Code:PAB15213
Catalog # 89-117-508
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to C-terminus of human SLIRP. |
Conjugate | DyLight 488 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. Do not freeze. |
Rabbit polyclonal antibody raised against synthetic peptide of SLIRP.
For Research Use Only
Protein Aliases: C14orf156; DC23; DC50; PD04872; SRA stem-loop-interacting RNA-binding protein, mitochondrial
Gene Aliases: C14orf156; DC23; DC50; PD04872; SLIRP
UniProt ID: (Human) Q9GZT3
Entrez Gene ID: (Human) 81892
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.