Novus Biologicals
Manufacturer Code:NBP180977
Catalog # NBP180977
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids:GRYEVELDAGVRTCPANPGAVCADSTSGLSRYQL |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Colon organic anion transporter; colon organic anion transporter OATP1 OATP4A1 OATPE OATP-E OATP-RP1 organic anion-transporting polypeptide E POAT SLC21A12 solute carrier family 21 (organic anion transporter) member 12 solute carrier family 21 member 12 solute carrier organic anion transporter family member 4A1 solute carrier organic anion transporter family member 4A1; OATP-E; OATP-RP1; OATP4A1; Organic anion transporter polypeptide-related protein 1; organic anion transporting polypeptide E; Organic anion-transporting polypeptide E; Sodium-independent organic anion transporter E; solute carrier family 21 (organic anion transporter), member 12; Solute carrier family 21 member 12; Solute carrier organic anion transporter family member 4A1; solute carrier organic anion transporter family, member 4A1
Gene Aliases: OATP-E; OATP1; OATP4A1; OATPE; OATPRP1; POAT; SLC21A12; SLCO4A1
UniProt ID: (Human) Q96BD0
Entrez Gene ID: (Human) 28231
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.