Abnova
Manufacturer Code:PAB22760
Catalog # 89-124-468
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein corresponding to amino acids of human SLCO4A1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against recombinant SLCO4A1.
For Research Use Only
Protein Aliases: Colon organic anion transporter; OATP-E; OATP-RP1; OATP1; OATP4A1; OATPE; OATPRP1; Organic anion transporter polypeptide-related protein 1; organic anion transporting polypeptide E; Organic anion-transporting polypeptide E; POAT; SLC21A12; Sodium-independent organic anion transporter E; solute carrier family 21 (organic anion transporter), member 12; Solute carrier family 21 member 12; Solute carrier organic anion transporter family member 4A1; solute carrier organic anion transporter family, member 4A1
Gene Aliases: OATP-E; OATP1; OATP4A1; OATPE; OATPRP1; POAT; SLC21A12; SLCO4A1
UniProt ID: (Human) Q96BD0
Entrez Gene ID: (Human) 28231
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.