Invitrogen
Manufacturer Code:PA584720
Catalog # PIPA584720
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human SLCO3A1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: OATP-D; OATP-D OATP3A1 SLC21A11; OATP-RP3; OATP3A1; Organic anion transporter polypeptide-related protein 3; Organic anion-transporting polypeptide D; PGE1 transporter; Sodium-independent organic anion transporter D; solute carrier family 21 (organic anion transporter), member 11; Solute carrier family 21 member 11; Solute carrier organic anion transporter family member 3A1; solute carrier organic anion transporter family, member 3A1
Gene Aliases: OATP-D; OATP-RP3; OATP3A1; OATPD; OATPRP3; SLC21A11; SLCO3A1
UniProt ID: (Human) Q9UIG8
Entrez Gene ID: (Human) 28232
Molecular Function:
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.