Invitrogen
Manufacturer Code:PA542457
Catalog # PIPA542457
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the middle region of human SLCO3A1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: androgen regulated protein 1; MJAM; OATP-D; OATP-RP3; OATP3A1; Organic anion transporter polypeptide-related protein 3; organic anion transporting polypeptide 3a1; Organic anion-transporting polypeptide D; PGE1 transporter; Prostaglandin transporter subtype 2; Sodium-independent organic anion transporter D; solute carrier family 21 (organic anion transporter), member 11; Solute carrier family 21 member 11; Solute carrier organic anion transporter family member 3A1; Solute carrier organic anion transporter family member 3A1 FLJ40478 OATP-D OATP3A1 SLC21A11 OATPD; solute carrier organic anion transporter family, member 3A1; zOatp3a2
Gene Aliases: 5830414C08Rik; Anr1; MJAM; OATP-D; OATP-RP3; OATP3A1; OATPD; OATPRP3; Pgt2; R75096; si:ch211-196l7.1; SLC21A11; SLCO3A1; wu:fa93e11
UniProt ID: (Bovine) Q8HYW2, (Human) Q9UIG8, (Mouse) Q8R3L5, (Rat) Q99N02
Entrez Gene ID: (Bovine) 407132, (Horse) 100070287, (Human) 28232, (Rabbit) 100341901, (Dog) 608833, (Mouse) 108116, (Rat) 140915, (Zebrafish) 569735
Molecular Function:
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.