Novus Biologicals
Manufacturer Code:NBP159678
Catalog # NBP159678
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptides corresponding to SLCO3A1(solute carrier organic anion transporter family member 3A1) The peptide sequence was selected from the middle region of SLCO3A1 (NP_037404). Peptide sequence MEIAVVAGFAAFLGKYLEQQFNLTTSSANQLLGMTAIPCACLGIFLGG |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: FLJ40478 OATP3A1OATP-RP3 OATP-DOATPRP3 OATPDSLC21A11 Organic anion transporter polypeptide-related protein 3 Organic anion-transporting polypeptide D PGE1 transporter Sodium-independent organic anion transporter D solute carrier family 21 (organic anion transporter) member 11 Solute carrier family 21 member 11 solute carrier organic anion transporter family member 3A1 solute carrier organic anion transporter family member 3A1; OATP-D; OATP-RP3; OATP3A1; Organic anion transporter polypeptide-related protein 3; Organic anion-transporting polypeptide D; PGE1 transporter; Sodium-independent organic anion transporter D; solute carrier family 21 (organic anion transporter), member 11; Solute carrier family 21 member 11; Solute carrier organic anion transporter family member 3A1; solute carrier organic anion transporter family, member 3A1
Gene Aliases: OATP-D; OATP-RP3; OATP3A1; OATPD; OATPRP3; SLC21A11; SLCO3A1
UniProt ID: (Human) Q9UIG8
Entrez Gene ID: (Human) 28232
Molecular Function:
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.