Abnova
Manufacturer Code:H00028232K
Catalog # 89-206-590
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Immunogen | A synthetic peptide of human SLCO3A1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Rabbit monoclonal antibody raised against a human SLCO3A1 peptide using ARM Technology.
For Research Use Only
Protein Aliases: FLJ40478 OATP-D OATP3A1 OATPD SLC21A11; OATP-D; OATP-RP3; OATP3A1; Organic anion transporter polypeptide-related protein 3; Organic anion-transporting polypeptide D; PGE1 transporter; Sodium-independent organic anion transporter D; solute carrier family 21 (organic anion transporter), member 11; Solute carrier family 21 member 11; Solute carrier organic anion transporter family member 3A1; solute carrier organic anion transporter family, member 3A1
Gene Aliases: OATP-D; OATP-RP3; OATP3A1; OATPD; OATPRP3; SLC21A11; SLCO3A1
UniProt ID: (Human) Q9UIG8
Entrez Gene ID: (Human) 28232
Molecular Function:
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.