Invitrogen
Manufacturer Code:PA587616
Catalog # PIPA587616
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fusion protein containing a sequence corresponding to amino acids 130-210 of human SLCO2B1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: OATP-B; OATP-RP2; Organic anion transporter B; Organic anion transporter polypeptide-related protein 2; solute carrier family 21 (organic anion transporter), member 9; Solute carrier family 21 member 9; Solute carrier organic anion transporter family member 2B1; solute carrier organic anion transporter family, member 2B1
Gene Aliases: AI060904; AI852653; KIAA0880; OATP-B; OATP2B1; OATPB; SLC21A9; SLCO2B1
UniProt ID: (Human) O94956, (Mouse) Q8BXB6
Entrez Gene ID: (Human) 11309, (Mouse) 101488
Molecular Function: transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.