Invitrogen
Manufacturer Code:PA542469
Catalog # PIPA542469
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human SLCO1C1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: BBB-specific anion transporter type 1; blood-brain barrier specific anion transporter; Blood-brain barrier-specific anion transporter 1; OAT-RP-5; OATP-14; OATP-F; OATP2; Organic anion transporter 2; Organic anion transporter F; Organic anion transporter polypeptide-related protein 5; organic anion transporting polypeptide 14; Organic anion-transporting polypeptide 14; solute carrier family 21 (organic anion transporter), member 14; Solute carrier family 21 member 14; Solute carrier organic anion transporter family member 1C1; Solute carrier organic anion transporter family member 1C1 OATP-F OATP1 OATP1C1 SLC21A14 OATPF OATP14 OATPRP5; solute carrier organic anion transporter family, member 1C1; Thyroxine transporter
Gene Aliases: Bsat1; OATP-14; OATP-F; OATP1; OATP14; OATP1C1; Oatp2; OATPF; OATPRP5; SLC21A14; SLCO1C1
UniProt ID: (Human) Q9NYB5, (Rat) Q9EPZ7, (Mouse) Q9ERB5
Entrez Gene ID: (Human) 53919, (Horse) 100068120, (Dog) 609603, (Bovine) 518047, (Rabbit) 100144344, (Rat) 84511, (Mouse) 58807
Molecular Function:
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.