Invitrogen
Manufacturer Code:PA5115919
Catalog # PIPA5115919
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human SLCO1C1 corresponding to a region within C-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: BBB-specific anion transporter type 1 blood-brain barrier specific anion transporter blood-brain barrier-specific anion transporter 1 Bsat1 OATP1 Oatp14 OATP-14 OATP1C1 Oatp2 OATPF OATP-F OATPRP5 OAT-RP-5 Organic anion transporter 2 organic anion transporter F organic anion transporter polypeptide-related protein 5 organic anion transporting polypeptide 14 Organic anion-transporting polypeptide 14 Slc21a14 SLCO1C1 solute carrier family 21 (organic anion transporter) member 14 solute carrier family 21 member 14 Solute carrier organic anion transporter family member 1C1 solute carrier organic anion transporter family member 1C1 thyroxine transporter; OAT-RP-5; OATP-14; OATP-F; Organic anion transporter F; Organic anion transporter polypeptide-related protein 5; organic anion transporting polypeptide 14; Organic anion-transporting polypeptide 14; solute carrier family 21 (organic anion transporter), member 14; Solute carrier family 21 member 14; Solute carrier organic anion transporter family member 1C1; solute carrier organic anion transporter family, member 1C1; Thyroxine transporter
Gene Aliases: OATP-F; OATP1; OATP14; OATP1C1; OATPF; OATPRP5; SLC21A14; SLCO1C1
UniProt ID: (Human) Q9NYB5
Entrez Gene ID: (Human) 53919
Molecular Function:
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.