Novus Biologicals
Manufacturer Code:NBP256965
Catalog # NBP256965
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:FLPNTLPKEGLETNADIIKNENEDKQKEEVKKEKYGITKDFLPFMKSLSCN |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: OATP-1; OATP-A; OATP1 OATP1A2member 3 OATP-AOATP-1 OATPSolute carrier family 21 member 3 Organic anion-transporting polypeptide 1 Sodium-independent organic anion transporter solute carrier organic anion transporter family member 1A2 solute carrier organic anion transporter family member 1A2; organic anion transporting polypeptide A; Organic anion-transporting polypeptide 1; Sodium-independent organic anion transporter; solute carrier family 21 (organic anion transporter), member 3; Solute carrier family 21 member 3; Solute carrier organic anion transporter family member 1A2; solute carrier organic anion transporter family, member 1A2
Gene Aliases: OATP; OATP-A; OATP1; OATP1A2; SLC21A3; SLCO1A2
UniProt ID: (Human) P46721
Entrez Gene ID: (Human) 6579
Molecular Function: transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.