Invitrogen
Manufacturer Code:PA569266
Catalog # PIPA569266
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide corresponding to a region of mouse SLC9A5 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: membrane protein; Na(+)/H(+) exchanger 5; NHE-5; sodium/hydrogen exchanger; Sodium/hydrogen exchanger 5; Sodium/hydrogen exchanger 5 Na(+)/H(+) exchanger 5 NHE-5 Solute carrier family 9 member 5 NHE5; solute carrier family 9 (sodium/hydrogen exchanger); solute carrier family 9 (sodium/hydrogen exchanger), isoform 5; solute carrier family 9 (sodium/hydrogen exchanger), member 5; Solute carrier family 9 member 5; solute carrier family 9, subfamily A (NHE5, cation proton antiporter 5), member 5
Gene Aliases: Gm696; NHE5; SLC9A5
UniProt ID: (Human) Q14940, (Rat) Q9Z0X2
Entrez Gene ID: (Bovine) 512686, (Dog) 609901, (Rabbit) 100349910, (Horse) 100053217, (Human) 6553, (Pig) 396753, (Mouse) 277973, (Rat) 192215
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.