Invitrogen
Manufacturer Code:PA5104162
Catalog # PIPA5104162
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human SLC9A5 corresponding to a region within C-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Gm696 Na(+)/H(+) exchanger 5 NHE5 NHE-5 SLC9A5 sodium/hydrogen exchanger 5 solute carrier family 9 (sodium/hydrogen exchanger) isoform 5 solute carrier family 9 (sodium/hydrogen exchanger) member 5 solute carrier family 9 member 5 solute carrier family 9 member A5 solute carrier family 9 subfamily A (NHE5 cation proton antiporter 5) member 5; Na(+)/H(+) exchanger 5; NHE-5; Sodium/hydrogen exchanger 5; solute carrier family 9 (sodium/hydrogen exchanger), isoform 5; solute carrier family 9 (sodium/hydrogen exchanger), member 5; Solute carrier family 9 member 5; solute carrier family 9, subfamily A (NHE5, cation proton antiporter 5), member 5
Gene Aliases: Gm696; NHE5; SLC9A5
UniProt ID: (Human) Q14940, (Rat) Q9Z0X2
Entrez Gene ID: (Human) 6553, (Mouse) 277973, (Rat) 192215
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.