Proteintech
Manufacturer Code:173981AP150UL
Catalog # 50-173-1551
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | SLC9A11 Fusion Protein Ag11351 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Na(+)/H(+) exchanger 11; Na(+)/H(+) exchanger 11 NHE 11 RP3 436N22.2 SLC9A11 Sodium/hydrogen exchanger 11; NHE-11; Sodium/hydrogen exchanger 11; Solute carrier family 9 member 11; Solute carrier family 9 member C2; solute carrier family 9, isoform 11; solute carrier family 9, member 11; solute carrier family 9, member C2 (putative)
Gene Aliases: RGD1565820; SLC9A11; SLC9C2
UniProt ID: (Human) Q5TAH2
Entrez Gene ID: (Human) 284525, (Rat) 364029
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.