Invitrogen
Manufacturer Code:PA577564
Catalog # PIPA577564
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide (C)PLEGKEVDESRRE corresponding to amino acid residues 303-315 of rat NCX-3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Na(+)/Ca(2+)-exchange protein 3; NCX-3; Sodium/calcium exchanger 3; sodium/calcium exchanger SLC8A3; solute carrier family 8 (sodium/calcium exchanger), member 3; Solute carrier family 8 member 3
Gene Aliases: AW742262; NCX3; SLC8A3
UniProt ID: (Human) P57103, (Mouse) S4R2P9, (Rat) P70549
Entrez Gene ID: (Human) 6547, (Mouse) 110893, (Rat) 140448
Molecular Function:
cation transporter
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.