Abnova
Manufacturer Code:PAB21082
Catalog # 89-122-578
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein corresponding to amino acids of human SLC8A3. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Protein Aliases: Na(+)/Ca(2+)-exchange protein 3; Sodium/calcium exchanger 3; sodium/calcium exchanger SLC8A3; solute carrier family 8 (sodium/calcium exchanger), member 3; Solute carrier family 8 member 3
Gene Aliases: NCX3; SLC8A3
UniProt ID: (Human) P57103
Entrez Gene ID: (Human) 6547
Molecular Function: cation transporter transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.