Invitrogen
Manufacturer Code:PA577563
Catalog # PIPA577563
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide (C)RVGDAQGMFEPDGG corresponding to amino acid residues 486-499 of rat NCX-2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Na(+)/Ca(2+)-exchange protein 2; Na+/Ca2+-exchanging protein Nac2; SLC8A2; sodium-calcium exchanger 2; Sodium/calcium exchanger 2; solute carrier family 8 (sodium-calcium exchanger), member 2; solute carrier family 8 (sodium/calcium exchanger), member 2; Solute carrier family 8 member 2
Gene Aliases: KIAA1087; NCX2; SLC8A2
UniProt ID: (Human) Q9UPR5, (Rat) P48768, (Mouse) Q8K596
Entrez Gene ID: (Human) 6543, (Rat) 140447, (Mouse) 110891
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.