Novus Biologicals
Manufacturer Code:NBP293474100UL
Catalog # NB028824
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 360-450 of human SLC8A2 (NP_055878.1). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: KIAA1087 Na(+)/Ca(2+)-exchange protein 2 Na+/Ca2+-exchanging protein Nac2 NCX2 sodium/calcium exchanger 2 sodium-calcium exchanger 2 solute carrier family 8 (sodium/calcium exchanger) member 2 solute carrier family 8 (sodium-calcium exchanger) member 2 solute carrier family 8 member 2; Na(+)/Ca(2+)-exchange protein 2; Na+/Ca2+-exchanging protein Nac2; sodium-calcium exchanger 2; Sodium/calcium exchanger 2; solute carrier family 8 (sodium-calcium exchanger), member 2; solute carrier family 8 (sodium/calcium exchanger), member 2; Solute carrier family 8 member 2
Gene Aliases: KIAA1087; NCX2; SLC8A2
UniProt ID: (Human) Q9UPR5
Entrez Gene ID: (Human) 6543
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.