Invitrogen
Manufacturer Code:PA5104159
Catalog # PIPA5104159
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human SLC8A1 corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AI852629 AV344025 CNC D930008O12Rik Na(+)/Ca(2+)-exchange protein 1 Na/Ca exchanger Na+/Ca++ exchanger Na+/Ca2+ exchanger Na-Ca exchanger Ncx Ncx1 SLC8A1 sodium/calcium exchanger 1 sodium/calcium exchanger isoform NaCa7 solute carrier family 8 (sodium/calcium exchanger) member 1 solute carrier family 8 member 1 solute carrier family 8 member A1 solute carrier family 8 member 1; Na(+)/Ca(2+)-exchange protein 1; Na+/Ca++ exchanger; Na+/Ca2+ exchanger; Na-Ca exchanger; Na/Ca exchanger; Sodium/calcium exchanger 1; sodium/calcium exchanger isoform NaCa7; solute carrier family 8 (sodium/calcium exchanger), member 1; Solute carrier family 8 member 1; solute carrier family 8, member 1
Gene Aliases: AI852629; AV344025; CNC; D930008O12Rik; Ncx; NCX1; SLC8A1
UniProt ID: (Human) P32418, (Mouse) P70414, (Rat) Q01728
Entrez Gene ID: (Human) 6546, (Mouse) 20541, (Rat) 29715
Molecular Function:
cation transporter
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.