Invitrogen
Manufacturer Code:PA5110394
Catalog # PIPA5110394
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 1-100 of human SLC7A9 (NP_055085.1). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: b(0+)AT b(0+)AT1 b(0+)-type amino acid transporter 1 BAT1 CSNU3 FLJ94301 Glycoprotein-associated amino acid transporter b0+AT1 SLC7A9 Solute carrier family 7 member 9; B(0,+)-type amino acid transporter 1; b(0,+)AT; b(0,+)AT1; b<o>, + amino acid transporter; b<o>, +AT; glycoprotein-associated amino acid transporter b0,+AT1; Glycoprotein-associated amino acid transporter b0,+AT1; solute carrier family 7 (amino acid transporter light chain, bo,+ system), member 9; solute carrier family 7 (cationic amino acid transporter, y+ system), member 9; solute carrier family 7 (glycoprotein-associated amino acid transporter light chain, bo,+ system), member 9; Solute carrier family 7 member 9
Gene Aliases: BAT1; CSNU3; SLC7A9
UniProt ID: (Human) P82251, (Rat) P82252, (Mouse) Q9QXA6
Entrez Gene ID: (Human) 11136, (Rat) 116726, (Mouse) 30962
Molecular Function: amino acid transporter transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.