Abnova
Manufacturer Code:PAB27931
Catalog # 89-129-453
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein corresponding to amino acids of human SLC7A9. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against recombinant SLC7A9.
For Research Use Only
Protein Aliases: b(0,+)-type amino acid transporter 1; b(0,+)AT; b(0,+)AT1; BAT1; CSNU3; FLJ94301; Glycoprotein-associated amino acid transporter b0,+AT1; solute carrier family 7 (amino acid transporter light chain, bo,+ system), member 9; solute carrier family 7 (cationic amino acid transporter, y+ system), member 9; solute carrier family 7 (glycoprotein-associated amino acid transporter light chain, bo,+ system), member 9; Solute carrier family 7 member 9
Gene Aliases: BAT1; CSNU3; SLC7A9
UniProt ID: (Human) P82251
Entrez Gene ID: (Human) 11136
Molecular Function: amino acid transporter transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.