OriGene
Manufacturer Code:TA500503
Catalog # 50-167-1636
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI5A9 |
Immunogen | Full-length protein expressed in 293T cell transfected with human SLC7A8 expression vector. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: hLAT2; integral membrane protein E16H; L-type amino acid transporter 2; Large neutral amino acids transporter small subunit 2; LAT2 LPI-PC1; solute carrier family 7 (amino acid transporter light chain, L system), member 8; solute carrier family 7 (amino acid transporter, L-type), member 8; solute carrier family 7 (cationic amino acid transporter, y+ system), member 8; Solute carrier family 7 member 8
Gene Aliases: LAT2; LPI-PC1; SLC7A8
UniProt ID: (Human) Q9UHI5
Entrez Gene ID: (Human) 23428
Molecular Function:
amino acid transporter
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.