Invitrogen
Manufacturer Code:PA542432
Catalog # PIPA542432
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the middle region of human SLC7A7 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: amino acid transporter SLC7A7 y+LAT1; Monocyte amino acid permease 2; MOP-2; Solute carrier family 7 (amino acid transporter light chain y+L system) member 7 LAT3 LPI Y+LAT1 y+LAT-1 MOP-2; solute carrier family 7 (amino acid transporter light chain, y+L system), member 7; solute carrier family 7 (cationic amino acid transporter, y+ system), member 7; Solute carrier family 7 member 7; y(+)L-type amino acid transporter 1; Y+L amino acid transporter 1; y+LAT-1; Y+LAT1
Gene Aliases: AI790233; LAT3; LPI; MOP-2; my+lat1; SLC7A7; y+LAT-1; Y+LAT1
UniProt ID: (Human) Q9UM01, (Rat) Q9R0S5, (Mouse) Q9Z1K8
Entrez Gene ID: (Bovine) 504220, (Dog) 490602, (Rabbit) 100356317, (Human) 9056, (Pig) 100037972, (Horse) 106782083, (Rat) 83509, (Guinea pig) 100732596, (Mouse) 20540
Molecular Function: amino acid transporter transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.