Invitrogen
Manufacturer Code:PA5113527
Catalog # PIPA5113527
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant Human Y+L amino acid transporter 1 protein (325-382AA) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AI790233 amino acid transporter SLC7A7 y+LAT1 BOS_10153 LAT3 LOC100356317 LPI Monocyte amino acid permease 2 MOP-2 my+lat1 Slc7a7 solute carrier family 7 (amino acid transporter light chain y+L system) member 7 solute carrier family 7 (cationic amino acid transporter y+ system) member 7 Solute carrier family 7 member 7 solute carrier family 7 member 7-like y(+)L-type amino acid transporter 1 Y+L amino acid transporter 1 Y+LAT1 y+LAT-1; Monocyte amino acid permease 2; MOP-2; solute carrier family 7 (amino acid transporter light chain, y+L system), member 7; solute carrier family 7 (cationic amino acid transporter, y+ system), member 7; Solute carrier family 7 member 7; y(+)L-type amino acid transporter 1; Y+L amino acid transporter 1; y+LAT-1; Y+LAT1
Gene Aliases: AI790233; LAT3; LPI; MOP-2; my+lat1; SLC7A7; y+LAT-1; Y+LAT1
UniProt ID: (Human) Q9UM01, (Mouse) Q9Z1K8
Entrez Gene ID: (Human) 9056, (Mouse) 20540
Molecular Function:
amino acid transporter
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.